missing translation for 'onlineSavingsMsg'
Learn More

Calcium Acetate, Monohydrate, Powder, ACS, 99%, Spectrum™ Chemical

Catalog No. 18609436
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18609436 50 kg Amber Glass Bottle
18609432 125 g Amber Glass Bottle
18609433 500 g Amber Glass Bottle
18609434 2.5 kg Amber Glass Bottle
18609435 12 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18609436 Supplier Spectrum Chemical Mfg Cor Supplier No. C106550KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5743-26-0
Molecular Formula C4H8CaO5
Molecular Weight (g/mol) 176.18
InChI Key XQKKWWCELHKGKB-UHFFFAOYSA-L
IUPAC Name calcium diacetate hydrate
SMILES O.[Ca++].CC([O-])=O.CC([O-])=O

Specifications

CAS Max % 1
Physical Form Powder, Needles or Crystals
Quantity 50 kg
Percent Purity 99%
Grade ACS/Reagent
Packaging Amber Glass Bottle
Chemical Name or Material Calcium Acetate, Monohydrate, Powder

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.