missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Acetate Monohydrate, ACS Reagent, ≥99%, Honeywell Fluka™
ACS Reagent,≥99.0%
$173.39 - $173.39
Chemical Identifiers
| CAS | 5743-26-0 |
|---|---|
| Molecular Formula | C4H8CaO5 |
| Molecular Weight (g/mol) | 176.181 |
| MDL Number | MFCD00150019 |
| InChI Key | XQKKWWCELHKGKB-UHFFFAOYSA-L |
| Synonym | calcium acetate monohydrate, calcium diacetate monohydrate, unii-7za48gim5h, calcium diacetate hydrate, acetic acid, calcium salt, monohydrate, calcium acetate, monohydrate, 7za48gim5h, calcium acetat, calcium acetate jan, acmc-1c5pu |
| PubChem CID | 82163 |
| ChEBI | CHEBI:59199 |
| IUPAC Name | calcium;diacetate;hydrate |
| SMILES | CC(=O)[O-].CC(=O)[O-].O.[Ca+2] |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
| 5743-26-0 | |
| 176.181 | |
| XQKKWWCELHKGKB-UHFFFAOYSA-L | |
| 82163 | |
| calcium;diacetate;hydrate |
| C4H8CaO5 | |
| MFCD00150019 | |
| calcium acetate monohydrate, calcium diacetate monohydrate, unii-7za48gim5h, calcium diacetate hydrate, acetic acid, calcium salt, monohydrate, calcium acetate, monohydrate, 7za48gim5h, calcium acetat, calcium acetate jan, acmc-1c5pu | |
| CHEBI:59199 | |
| CC(=O)[O-].CC(=O)[O-].O.[Ca+2] |
Specifications
| White | |
| 100 g | |
| Ca(CH3CO2)2 · H2O | |
| NONH for all modes of transport | |
| XQKKWWCELHKGKB-UHFFFAOYSA-L | |
| calcium;diacetate;hydrate | |
| 82163 | |
| 176.18g/mol | |
| Poly Bottle | |
| Calcium acetate monohydrate |
| 5743-26-0 | |
| C4H8CaO5 | |
| MFCD00150019 | |
| calcium acetate monohydrate, calcium diacetate monohydrate, unii-7za48gim5h, calcium diacetate hydrate, acetic acid, calcium salt, monohydrate, calcium acetate, monohydrate, 7za48gim5h, calcium acetat, calcium acetate jan, acmc-1c5pu | |
| CC(=O)[O-].CC(=O)[O-].O.[Ca+2] | |
| 176.181 | |
| CHEBI:59199 | |
| ≥99% | |
| Powder |
Safety and Handling
RTECSNumber : 200-540-9