missing translation for 'onlineSavingsMsg'
Learn More

Calcium Acetate, EP, USP, 99-100.5%, Spectrum™ Chemical

Catalog No. p-7157107
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Amber Glass Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18601206 100 g Amber Glass Bottle
18601203 500 g Amber Glass Bottle
18601204 2.5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18601206 Supplier Spectrum Chemical Mfg Cor Supplier No. C1067100GM
Only null left

C1067, 62-54-4, C4H6CaO4

Spectrum™ Chemical Calcium Acetate, USP, EP is used to prevent high blood phosphate levels in patients who are on dialysis due to severe kidney disease. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 62-54-4
Molecular Formula C4H6CaO4
Molecular Weight (g/mol) 158.17
MDL Number MFCD00012448
InChI Key VSGNNIFQASZAOI-UHFFFAOYSA-L
IUPAC Name calcium diacetate
SMILES [Ca++].CC([O-])=O.CC([O-])=O

Specifications

pH 7.2-8.2
CAS Max % 100%
Quantity 100 g
Percent Purity 99 to 100.5%
Grade USP
Packaging Amber Glass Bottle