missing translation for 'onlineSavingsMsg'
Learn More

Calcium, 99%, granular, Thermo Scientific Chemicals

Catalog No. p-222775
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Plastic Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC201180050 5 g Plastic Bottle
AC201181000 100 g Plastic Bottle
AC201185000 500 g Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC201180050 Supplier Thermo Scientific Chemicals Supplier No. 201180050
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-70-2
Molecular Formula Ca
Molecular Weight (g/mol) 40.08
MDL Number MFCD00010897 MFCD00085314
InChI Key OYPRJOBELJOOCE-UHFFFAOYSA-N
Synonym unii-sy7q814vup, hsdb 273, sy7q814vup, compounds, elemental, calcium, elemental, kalzium, atom, ingot, chelate
PubChem CID 5460341
ChEBI CHEBI:29320
IUPAC Name calcium
SMILES [Ca]

Specifications

Color Gray
Melting Point 845.0°C
Boiling Point 1484.0°C
Quantity 5 g
Packaging Plastic Bottle
Form Granular
Assay Percent Range 99%
Linear Formula Ca
Fieser 05,89; 09,1634; 14,71
Merck Index 15, 1648
Formula Weight 40.07
Percent Purity 99%
Chemical Name or Material Calcium, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
In contact with water releases flammable gases.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Handle under inert gas. Protect from moisture.
Keep away from any possible contact with water, because of violent reaction and possible flash fire.
Brush off loose particles from skin. Immerse in cool water/wrap in wet bandages.
In case of fire: Use CO2, dry chemical, or foam.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 231-179-5
TSCA TSCA

RUO – Research Use Only