missing translation for 'onlineSavingsMsg'
Learn More

Calcium 3-Hydroxy-3-methylbutyrate Hydrate 97.0+%, TCI America™

Catalog No. H091725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H091725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H091725G Supplier TCI America Supplier No. H091725G
Only null left

Chemical Identifiers

CAS 135236-72-5
Molecular Formula C10H18CaO6
Molecular Weight (g/mol) 274.33
MDL Number MFCD01318562
InChI Key WLJUMPWVUPNXMF-UHFFFAOYSA-L
Synonym calcium 3-hydroxy-3-methyl butyrate
PubChem CID 131847069
IUPAC Name calcium bis(3-hydroxy-3-methylbutanoate)
SMILES [Ca++].CC(C)(O)CC([O-])=O.CC(C)(O)CC([O-])=O

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 274.33
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Calcium 3-Hydroxy-3-methylbutyrate Hydrate