missing translation for 'onlineSavingsMsg'
Learn More

Calcium 3-hydroxy-3-methylbutyrate hydrate, 97+%

Catalog No. p-7052176
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4272118 50 g
AA4272130 250 g
AA42721A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4272118 Supplier Thermo Scientific Chemicals Supplier No. 04272118
Only null left

CAS: 135236-72-5 | C10H18CaO6 | 274.33 g/mol

Calcium 3-hydroxy-3-methylbutyrate hydrate is a nutritional supplement, which represents a calcium salt of beta-hydroxy beta-methylbutyrate (HMB). The latter is a leucine metabolite produced in the body through oxidation of the ketoacid of leucine (ketoisocaproate, KIC). Because leucine plays an important regulatory role in protein metabolism and due to the conversion of leucine to KIC, several researchers have presumed that KIC is the active ingredient responsible for most of the beneficial effects of leucine on protein metabolism.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 135236-72-5
Molecular Formula C10H18CaO6
Molecular Weight (g/mol) 274.33
MDL Number MFCD01318562
InChI Key WLJUMPWVUPNXMF-UHFFFAOYSA-L
Synonym calcium 3-hydroxy-3-methyl butyrate
PubChem CID 131847069
SMILES [Ca++].CC(C)(O)CC([O-])=O.CC(C)(O)CC([O-])=O

Specifications

Color White
Quantity 50 g
Solubility Information Soluble in water.
IUPAC Name calcium bis(3-hydroxy-3-methylbutanoate)
Formula Weight 274.33 (Anhydrous)
Percent Purity ≥97%
Physical Form Powder
Chemical Name or Material Calcium 3-hydroxy-3-methylbutyrate hydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only