Learn More
Thermo Scientific Chemicals Calcitonin, human, 96%
For cellular and molecular biology applications
$517.23 - $706.12
Specifications
| Species | Human |
|---|---|
| Purification Method | HPLC |
| Purity | ≥97% |
| Common Name | Human C carcinoma |
| Content And Storage | -30°C to -10°C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAJ63192EXD
|
Thermo Scientific Chemicals
J63192EXD |
0.5 mg |
Each for $517.23
|
|
|||||
|
AAJ63192MCR
|
Thermo Scientific Chemicals
J63192MCR |
1 mg |
Each for $706.12
|
|
|||||
Description
Decreases blood calcium and phosphate due to inhibition of resorption by osteoblasts and osteocytes. A carrier peptide that can be used to internalize fusion proteins
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Human | |
| ≥97% | |
| -30°C to -10°C | |
| Decreases blood calcium and phosphate due to inhibition of resorption by osteoblasts and osteocytes. A carrier peptide that can be used to internalize fusion proteins | |
| Calcitonin, human, ≥97% | |
| Calcitonin M |
| HPLC | |
| Human C carcinoma | |
| Powder | |
| 3417.90 | |
| RUO - research use only |
RUO – Research Use Only