missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Calcitonin Gene Related Peptide II, Human

Catalog No. AAJ64037EXD
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ64037EXD 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ64037EXD Supplier Thermo Scientific Chemicals Supplier No. J64037EXD

 

Calcitonin Gene Related Peptide II is a member of the calcitonin gene family used to study CGRP receptor specificity and function and CGRP signaling. Potent, long-lasting vasodilator; activation of CGRP receptors on pancreatic β-cells increases plasma levels of pancreatic enzymes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Species Human
Purification Method HPLC
Content And Storage -30°C to -10°C
Format Solid
Molecular Weight (g/mol) 3793.35
Product Type Calcitonin Gene Related Peptide II, human
Quantity 0.5 mg
Regulatory Status RUO - research use only

RUO – Research Use Only