missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Calcitonin Gene Related Peptide, Human
Description
Calcitonin Gene Related Peptide is used as a potent, long-lasting vasodilator, activation of CGRP receptors on pancreatic β-cells increases plasma levels of pancreatic enzymes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Species | Human |
| Purification Method | HPLC |
| Content And Storage | -30°C to -10°C |
| Format | Solid |
| Molecular Weight (g/mol) | 3789.3 |
| Product Type | Calcitonin Gene Related Peptide, human |
| Quantity | 0.5 mg |
| Regulatory Status | RUO - research use only |
| Sequence | CGRP, human |
RUO – Research Use Only