missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Calcined petroleum coke powder, crystalline, -325 mesh, 99%

CAS: 64743-05-1

$72.38 - $146.68

Specifications

CAS 64743-05-1
Density 2.1
Physical Form Solid
Synonym Coke (petroleum), calcined;
Percent Purity 99%
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4727514
View Documents
Thermo Scientific™
4727514
25 g
Each for $72.38
Only null left
 
AA4727530
View Documents
Thermo Scientific™
4727530
250 g
Each for $93.40
Only null left
 
AA47275A1
View Documents
Thermo Scientific™
47275A1
1 kg
Each for $146.68
Only null left
 
Description

Description

Specifications

Specifications

64743-05-1
Solid
99%
2.1
Coke (petroleum), calcined;
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'