missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Calcined petroleum coke powder, crystalline, -325 mesh, 99%

Catalog No. AA4727514
Change view
Click to view available options
Quantity:
25 g
250 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4727514 25 g
AA4727530 250 g
AA47275A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4727514 Supplier Thermo Scientific™ Supplier No. 4727514

CAS: 64743-05-1

Specifications

CAS 64743-05-1
Density 2.1
Quantity 25 g
Physical Form Solid
Synonym Coke (petroleum), calcined;
Percent Purity 99%