missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Calcined petroleum coke powder, -20+80 mesh

Catalog No. p-7237366
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4728730 250 g
AA47287A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4728730 Supplier Thermo Scientific™ Supplier No. 4728730
Only null left

CAS: 64743-05-1

Chemical Identifiers

CAS 64743-05-1
Synonym Coke (petroleum), calcined;

Specifications

Density 2.1
Quantity 250 g
Physical Form Solid
Percent Purity 99.9%
ShelfLife 5 Years
EINECSNumber 265-210-9
RTECSNumber GH0359000