missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calciferol 98.0+%, TCI America™
$131.00 - $131.00
Chemical Identifiers
| CAS | 50-14-6 |
|---|---|
| Molecular Formula | C28H44O |
| Molecular Weight (g/mol) | 396.66 |
| MDL Number | MFCD00166988,MFCD11656674 |
| InChI Key | MECHNRXZTMCUDQ-VLOQVYPSSA-N |
| Synonym | ergocalciferol, vitamin d2, calciferol, viosterol, oleovitamin d2, ercalciol, ergorone, detalup, condocaps, crystallina |
| PubChem CID | 134129658 |
| IUPAC Name | (1S,3E)-3-{2-[(1R,3aS,4E,7aR)-1-[(2R,3E,5R)-5,6-dimethylhept-3-en-2-yl]-7a-methyl-octahydro-1H-inden-4-ylidene]ethylidene}-4-methylidenecyclohexan-1-ol |
| SMILES | CC(C)[C@@H](C)\C=C\[C@@H](C)[C@H]1CC[C@H]2\C(CCC[C@]12C)=C\C=C1/C[C@@H](O)CCC1=C |
Chemical Identifiers
| 50-14-6 | |
| 396.66 | |
| MECHNRXZTMCUDQ-VLOQVYPSSA-N | |
| 134129658 | |
| CC(C)[C@@H](C)\C=C\[C@@H](C)[C@H]1CC[C@H]2\C(CCC[C@]12C)=C\C=C1/C[C@@H](O)CCC1=C |
| C28H44O | |
| MFCD00166988,MFCD11656674 | |
| ergocalciferol, vitamin d2, calciferol, viosterol, oleovitamin d2, ercalciol, ergorone, detalup, condocaps, crystallina | |
| (1S,3E)-3-{2-[(1R,3aS,4E,7aR)-1-[(2R,3E,5R)-5,6-dimethylhept-3-en-2-yl]-7a-methyl-octahydro-1H-inden-4-ylidene]ethylidene}-4-methylidenecyclohexan-1-ol |
Specifications
| 50-14-6 | |
| White | |
| MFCD00166988,MFCD11656674 | |
| 2811 | |
| MECHNRXZTMCUDQ-VLOQVYPSSA-N | |
| (1S,3E)-3-{2-[(1R,3aS,4E,7aR)-1-[(2R,3E,5R)-5,6-dimethylhept-3-en-2-yl]-7a-methyl-octahydro-1H-inden-4-ylidene]ethylidene}-4-methylidenecyclohexan-1-ol | |
| 134129658 | |
| ≥98.0% (HPLC) | |
| Calciferol |
| 114°C | |
| C28H44O | |
| 1 g | |
| ergocalciferol, vitamin d2, calciferol, viosterol, oleovitamin d2, ercalciol, ergorone, detalup, condocaps, crystallina | |
| CC(C)[C@@H](C)\C=C\[C@@H](C)[C@H]1CC[C@H]2\C(CCC[C@]12C)=C\C=C1/C[C@@H](O)CCC1=C | |
| 396.66 | |
| 396.66 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : KE1050000
TSCA : Yes
Recommended Storage : Refrigerator