missing translation for 'onlineSavingsMsg'
Learn More

Calciferol, 97%, Thermo Scientific Chemicals

Catalog No. AAJ6216306
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6216306 5 g
AAJ6216303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6216306 Supplier Thermo Scientific Chemicals Supplier No. J6216306

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 50-14-6 | Molecular Formula: C28H44O | Molecular Weight: 396.66 g/mol

Calciferol has a strong inhibitory effect against bladder tumor promotion by sodium saccharin and induces cell differentiation in leukemia cells. Calciferol has also been shown to be an inhibitor of DNA polymerase. Calciferol with calcium is used to treat or prevent bone loss (osteoporosis). Calciferol is also used with other medications to treat low levels of calcium or phosphate caused by certain disorders (such as hypoparathyroidism, pseudohypoparathyroidism, familial hypophosphatemia). It is often used in kidney disease to keep calcium levels normal and allow normal bone growth.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in ethanol (80 mg/ml at 25°C), water (<1mg/mL at 25°C), methanol, DMF (∼20 mg/ml), and DMSO (20 mg/ml at 25°C).

Notes
Store at 4°C. Incompatible with heat and oxidizing agents.

Chemical Identifiers

CAS 50-14-6
Molecular Formula C28H44O
Molecular Weight (g/mol) 396.66
MDL Number MFCD00166988,MFCD11656674
InChI Key MECHNRXZTMCUDQ-VLOQVYPSSA-N
Synonym ergocalciferol, vitamin d2, calciferol, viosterol, oleovitamin d2, ercalciol, ergorone, detalup, condocaps, crystallina
PubChem CID 134129658
IUPAC Name (1S)-3-[2-[(1S,7aR)-1-[(2R,5R)-5,6-dimethylhept-3-en-2-yl]-7a-methyl-2,3,3a,5,6,7-hexahydro-1H-inden-4-ylidene]ethylidene]-4-methylidenecyclohexan-1-ol
SMILES CC(C)[C@@H](C)\C=C\[C@@H](C)[C@H]1CC[C@H]2\C(CCC[C@]12C)=C\C=C1/C[C@@H](O)CCC1=C

Specifications

Melting Point 118°C to 119°C
Color White
Quantity 5 g
UN Number UN2811
Beilstein 1916682
Merck Index 14,10018
Solubility Information Soluble in ethanol (80mg/ml at 25°C),water (<1mg/ml at 25°C),methanol,DMF (∽20mg/ml),and DMSO (20mg/ml at 25°C).
Optical Rotation +110° (c=1.5 in Ethanol)
Formula Weight 396.66
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Calciferol
Show More Show Less
Hazard Category H301+H311-H330-H372
Hazard Statement GHS H Statement
H301-H311-H330-H372
Toxic if swallowed.
Toxic in contact with skin.
Fatal if inhaled.
Causes damage to organs through prolonged or repeated exposure.
Precautionary Statement P260-P264b-P270-P271-P280-P284-P301+P310-P302+P352-P304+P340-P310-P312-P314-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 200-014-9
RTECSNumber KE1050000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only