missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Total Antioxidant Status Assay Kit

Catalog No. 6157001KIT
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6157001KIT 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6157001KIT Supplier MilliporeSigma™ Supplier No. 6157001KIT

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Substrate, Chromogen, Standard, Buffer, and user protocol

Used for measurement of total antioxidant status.

EMD Millipore Calbiochem™ Total Antioxidant Status Assay Kit is colorimetric assay suitable for performing a minimum of 50 assays.

  • Colorimetric detection
  • Format: Cuvette, Sufficient for 50 Tests
  • Assay Range: 0 to 25nmol/10μl plasma
  • Sample Type: Serum, plasma, tissue extracts, plant extracts, beverages
  • Packaging: Glass Bottle

Specifications

Detection Method Colorimetric
Quantity 1 Kit
For Use With (Application) Assay antioxidant levels in serum or plasma samples
Content And Storage 2° to 8°C