missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ TMB, Soluble Substrates for AP and HRP

Catalog No. 613544100ML
Change view
Click to view available options
Volume (Metric):
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
613544100ML 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 613544100ML Supplier MilliporeSigma™ Supplier No. 613544100ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Peroxidase substrate that produces blue soluble end product that can be read at 370nm or 650nm. Calbiochem™ TMB, Soluble Substrates for AP and HRP use of stop solution enhances sensitivity two- to four-fold and results in yellow solution that can be read at 450nm.

  • CAS number: 54827-17-7
  • Form: Liquid, clear to very light blue
  • Preservative: None
  • Special instructions: Protect from exposure to direct sunlight. Discard if the solution turns blue or turbid. The reagent is stable at room temperature, but we recommend storage in the refrigerator for longer shelf life. Warm to assay temperature before use.

Specifications

Product Type TMB, Soluble
Formulation Single component system containing 1.46mM TMB, 2.21mM H₂O₂ in a proprietary solvent
For Use With (Application) ELISA
pH 3.1 ± 0.5
Storage Requirements Protect from light. Ok to freeze.
Volume (Metric) 100mL
Form Liquid