missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Thiol Fluorescent Probe IV
Description
EMD Millipore Calbiochem™ Thiol Fluorescent Probe IV is cell-permeable, coumarin cis-acrylate derivative.
- Displays high reactivity toward 2-ME, GSH, Cysteine, but not non-thiol-containing GSSG or Cystine
- Fluorescently labels cellular thiols in intact HEK293 cells
- Detects glutathione reductase activity in cell-free enzymatic assays
- Assay: ≥98% by HPLC
- CAS: 1173888-41-9
- Molecular Formula: C14H11NO6
Specifications
Specifications
| Description | Thiol Fluorescent Probe IV |
| Color | Brown |
| Concentration | 10 mg/mL |
| Quantity | 10 mg |
| Storage Requirements | −20°C |
| Product Type | Fluorescent Dye |