missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Superoxide Dismutase Assay Kit II

Catalog No. 5746011KIT
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5746011KIT 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 5746011KIT Supplier MilliporeSigma™ Supplier No. 5746011KIT

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Assay Buffer, Sample Buffer, Radical Detector, SOD Standard, Xanthine Oxidase, 96-Well Plate, Plate Cover, and user protocol

Used for measurement of three types (Cu/Zn, Mn, Fe) of SOD.

EMD Millipore Calbiochem™ Superoxide Dismutase Assay Kit II is sensitive spectrophotometric assay.

  • Positive control: Bovine erythrocyte SOD (Cu/Zn)
  • Unit of Definition: Amount of enzyme needed to exhibit 50% dismutation of superoxide radical
  • Format: 96 well plate
  • TotalAssayTime: 1.5 hr
  • Sufficient for 96 Tests
  • Assay Range: 0.025 to 0.25U/mL SOD
  • Sample Type: Plasma, serum, urine, tissue culture medium tissue homogenates
  • Packaging: Glass Bottle

Specifications

Detection Method Colorimetric
Quantity 1 Kit
Content And Storage −20°C