missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Calbiochem™ RAPIDstain™ Reagent
Description
EMD Millipore Calbiochem™ RAPIDstain™ Reagent is ultrasensitive Coomassie-based reagent.
- No mixing or preparation is involved
- No fixation step is needed
- No destaining is required
- RAPIDstain™ Reagent stains only protein, within 5 to 10 min.
- Leaves crystal-clear background with high resolution
- Note: 1L is sufficient to stain up to 20 minigels or 10 standard (16 x 18cm) gels
- Sensitivity 4 to 8ng BSA
Specifications
Specifications
| Description | RAPID stain Reagent |
| Color | Blue |
| Quantity | 1 L |
| Storage Requirements | 15° to 30°C |
| Product Type | Protein Stain |