Learn More
MilliporeSigma™ Calbiochem™ Normal Goat IgG
Catalog No. ni02100ug
Change view
Click to view available options
:
Normal Goat IgG
Catalog No.
NI02100UG Normal Goat IgG
1 options
Catalog No. NI02100UG Supplier MilliporeSigma™ Supplier No. NI02100UG
Only null left
Used in flow cytometry, immunoblotting, Immunofluorescence, immunohistochemistry, immunoprecipitation.
  • Negative control in parallel with specific goat IgG-type primary antibodies
Specifications
Quantity 100 μg
Content And Storage 2°C to 8°C