missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Nitric Oxide Assay Kit (Fluorometric)

Catalog No. 4826551KIT
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4826551KIT 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4826551KIT Supplier MilliporeSigma™ Supplier No. 4826551KIT

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Assay Buffer, Nitrate Reductase, Enzyme Cofactors, Nitrate Standard, Nitrite Standard, DAN Reagent, NaOH, Microtiter Plates, Plate Covers, and user protocol

Used for quantitative measurement of total nitric oxide.

EMD Millipore Calbiochem™ Nitric Oxide Assay Kit (Fluorometric) is based on enzymatic conversion of nitrate to nitrite by nitrate reductase.

  • Fluorometric detection
  • Displays 50 fold increased sensitivity
  • Format: 96 well plate
  • TotalAssayTime: 3 to 5 hr
  • Sufficient for 192 to 288 Tests
  • Sample Type: Aqueous solutions

Specifications

Detection Method Fluorometric
Quantity 1 Kit
Content And Storage −20°C