missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ N-Acetyllactosamine

Catalog No. 34525020MG
Change view
Click to view available options
Quantity:
20 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34525020MG 20 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 34525020MG Supplier MilliporeSigma™ Supplier No. 34525020MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Useful as lectin inhibitory sugar

  • ;Acts as substrate for galactosidases, fucosyltransferases, and sialyltransferases

Specifications

Color White to Off-white
Content And Storage 2°C to 8°C
Quantity 20 mg
Product Type N-Acetyllactosamine
Form Solid

For Research use Only