missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Mitochondrial Division Inhibitor, mdivi-1
Description
Packaged Under Inert Gas
Specifications
Specifications
| Color | White |
| Description | 95% by HPLC |
| CAS | 338967-87-6 |
| Molecular Formula | C15H10Cl2N2O2S |
| Formula Weight | 353.2g/mol |
| Solubility | DMSO |
| Content And Storage | Protect from light |
| Form | Solid |
| Quantity | 10 mg |