missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Marimastat

Catalog No. 4442895MG
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4442895MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4442895MG Supplier MilliporeSigma™ Supplier No. 4442895MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Used in studying involvement of MMPs in various cellular and pathological processes both in vitro and in vivo

  • Orally active peptidyl hydroxamate-based broad-spectrum MMP inhibitor

Specifications

Color White
Content And Storage 2°C to 8°C
Quantity 5 mg
Product Type Marimastat
Form Solid

For Research use Only