missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Inosine

Catalog No. 406050GM
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
406050GM 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 406050GM Supplier MilliporeSigma™ Supplier No. 406050G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Acts as potent coronary vasodilator

  • Metabolite of adenosine
  • Exhibits positive inotropic action on heart muscle
  • Can antagonize ouabain-induced arrhythmias

Specifications

Format White solid
Product Type Inosine
Content And Storage 15°C to 30°C
Purity ≥99% by assay
Quantity 50 mg

For Research Use Only