missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Human IgG, Fab Fragment, Plasma
Specifications
Specifications
| Formulation | In 150mM NaCl, 20mM sodium phosphate buffer, pH 7.4 |
| Quantity | 2 mg |
| Test Specificity | Negative for HBsAg and for HIV and HCV antibodies |
| Content And Storage | -20°C |
| Form | Liquid |
For Research Use Only