missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Human IgD, Myeloma Plasma

Catalog No. 401164100UG
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
401164100UG 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 401164100UG Supplier MilliporeSigma™ Supplier No. 401164100UG

Native IgD from human myeloma plasma

Specifications

Formulation In 150mM NaCl, 10mM TRIS-HCl, 10 mM EACA, pH 8.0
Quantity 100 μg
Test Specificity Negative for HBsAg and for HIV and HCV antibodies
Content And Storage -20°C
Form Liquid

For Research Use Only