missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Human IgA, Plasma
Specifications
Specifications
| Formulation | In 100mM NaCl, 100mM Tris-HCl, pH 8 |
| Quantity | 2 mg |
| Test Specificity | Nonreactive for HBsAg and for HCV, HBc, HIV-1, and HIV-2 antibodies |
| Content And Storage | -20°C |
| Form | Liquid |
For Research Use Only