missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Calbiochem™ HIV Reverse Transcriptase, Recombinant E. coli
Provides an efficient target for antiviral agents.
Supplier: MilliporeSigma™ 382129500U
Description
EMD Millipore Calbiochem™ HIV Reverse Transcriptase, Recombinant E. coli catalyzes error-prone synsis from DNA RNA templates.
- Expressed in E. coli
- Activity ≥10,000U/mL
- One unit is defined as the amount of enzyme that will incorporate 1.0nmol of labeled dTTP into acid-insoluble material in 20 min. at 37°C, pH 8.3
- Specific Activity ≥5,000U/mg protein
- Contaminants Nuclease: None Detected
Recommended Storage
−20°C
Specifications
HIV Reverse Transcriptase, Recombinant E. coli | |
HIV Reverse Transcriptase |