missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ H-Gly-Pro-Arg-Pro-OH

Catalog No. 0334000125M
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
0334000125M 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 0334000125M Supplier MilliporeSigma™ Supplier No. 0334000125MG

Inhibitor of fibrinogen aggregation and fibrin polymerization

Specifications

Purity ≥98% by HPLC
Content And Storage -20°C
Format Lyophilized
Formulation Supplied as an acetate salt
Product Type Peptide
Quantity 25 mg
Sequence H-Gly-Pro-Arg-Pro-OH

For Research Use Only