missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ H-Arg-Gly-Asp-Ser-OH

Catalog No. 0334000225M
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
0334000225M 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 0334000225M Supplier MilliporeSigma™ Supplier No. 0334000225MG

Shown to inhibit fibronectin function by binding to platelet-binding sites and to induce apoptosis in osteoblasts

Specifications

Purity ≥97% by HPLC
Color White to Off-white
Content And Storage -20°C
Format Powder
Formulation Supplied as a trifluoroacetate salt
Product Type Peptide
Quantity 25 mg
Sequence H-Arg-Gly-Asp-Ser-OH

For Research Use Only