missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Guanosine 5'-Triphosphate, Disodium Salt

Catalog No. p-5533227
Change view
Click to view available options
Quantity:
50 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37170150MG 50 mg
371701250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 37170150MG Supplier MilliporeSigma™ Supplier No. 37170150MG
Only null left

For biochemical applications

Specifications

Format White solid
Product Type Guanosine 5′-Triphosphate, Disodium Salt
Content And Storage -20°C
Purity ≥90% by HPLC
Quantity 50 mg

For Research Use Only