missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Gluconolactone

Oxidation product of glucose by glucose oxidase.

Supplier:  MilliporeSigma™ 347425500GM

Catalog No. 347425500GM


Only null left

Description

Description

  • Produced from glucose by glucose oxidase on oxidation
  • Assay: ≥99% by Acidimetric Analysis
  • CAS: 90-80-2
  • Molecular Formula: C6H10O6
  • Color: White
Specifications

Specifications

15°C to 30°C
Gluconolactone
500 g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions