Learn More
MilliporeSigma™ CalBiochem™ FragEL™ DNA Fragmentation Detection Kit, Fluorescent -TdT Enzyme
Catalog No. QIA391EA
Change view
Click to view available options
Quantity:
50 Tests/Kit
Catalog No. Quantity
QIA391EA 50 Tests/Kit
1 options
Catalog No. QIA391EA Supplier MilliporeSigma™ Supplier No. QIA391EA
Includes
Counterstain, Proteinase K, Equilibration and Labeling Buffers, Klenow Enzyme, Stop buffer, Blocking Buffer, 50X Conjugate, DAB, Positive and Negative Control Slides, H2O2/Urea Tablets, Methyl Green Counterstain, and user protocol.
Used as blocking agent in Western blots, dot blots ELISA.
  • Unique modified milk protein
  • Semi-clear 2 to 5% solution
  • High blocking efficiency
  • Blocks within 15 to 20 min.
  • Produces clear background
  • Does not inhibit peroxidase
  • Packaging: Fibre Case
Specifications
Quantity 50 Tests/Kit
Storage Requirements −20°C
Product Type Fluorescent DNA Fragmentation Detection Kit