missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Formyl-Met-Leu-Phe-OH

Catalog No. 0522250025M
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
0522250025M 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 0522250025M Supplier MilliporeSigma™ Supplier No. 0522250025MG

Chemotactic peptide for polymorphonuclear leukocytes

Specifications

Purity ≥99% by HPLC
Color White to Off-white
Content And Storage -20°C
Format Lyophilized Solid
Product Type Formyl-Met-Leu-Phe-OH
Quantity 25 mg
Sequence Formyl-Met-Leu-Phe-OH

For Research Use Only