missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ FluorSave™ Reagent

Catalog No. 34578920ML
Change view
Click to view available options
Volume (Metric):
20 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
34578920ML 20 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 34578920ML Supplier MilliporeSigma™ Supplier No. 34578920ML

Used in conjunction with fluorescent tracers.

EMD Millipore Calbiochem™ FluorSave™ Reagent is aqueous mounting medium for use with phycobiliprotein tracers.

  • Used with Fluorescent tracers such as fluorescein, rhodamine, phycobiliprotein probes
  • Preserves fluorescence-stained tissue sections
  • Specially formulated without glycerol

Specifications

Product Type FluorSave Reagent
Packaging Glass Bottle
Content And Storage 15° to 30°C
Volume (Metric) 20 mL