missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Calbiochem™ D(+)Cellobiose
Description
Substrate for β-glucosidase
Specifications
Specifications
| CAS | 528-50-7 |
| Color | White |
| Molecular Formula | C12H22O11 |
| Quantity | 25 g |
| Synonym | Glcβ1,4Glc, 4-O-β-D-Glucopyranosyl-D-glucose |
| Molecular Weight (g/mol) | 342.3 |
| Percent Purity | ≥98% (HPLC) |
| Physical Form | Crystalline Powder |