missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Congo Red, High Purity

Useful for staining visual detection of amyloid deposits.

Supplier:  MilliporeSigma™ 234610500MG

Catalog No. 234610500MG


Only null left

Description

Description

EMD Millipore Calbiochem™ Congo Red, High Purity is an amyloidophylic dye that specifically stains stacked β sheet aggregates.

  • Does not bind to non-polymerized amyloid peptide
  • Inhibits accumulation of scrapie-associated, protease-resistant isoform of protein PrP without affecting metabolism of normal isoform
  • Inhibits Pot1 binding to telomeric DNA
  • Not ATP competitive, reversible, cell permeable
  • Assay: ≥75% (dye content)
  • CAS: 573-58-0
  • Molecular Formula: C32H22N6O6S2 · 2Na
Specifications

Specifications

Congo Red, High Purity
Stackedβsheet aggregates
15° to 30°C
Red
500 mg
Amyloidophylic dye
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

ShelfLife : Stock solutions are Stable for up to 1 month at room temperature (20°C).

SDS
Documents

Documents

Promotions

Promotions