missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Cdk1 Inhibitor IV, RO-3306

Catalog No. 2176995mg
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
5 mg
Catalog No. Quantity
2176995MG 5 mg
1 options

Catalog No. 2176995MG

Supplier: MilliporeSigma™ 2176995MG

Only null left

RO-3306 is a cell-permeable, potent and ATP-competitive inhibitor of Cdk1 (Ki = 35 nM and 110 nM for Cdk1/B1 and Cdk1/A, respectively).

  • Packaged Under Inert Gas
  • Cell Permeable

Specifications

Color Brown
Description 95% by HPLC
Target Proteins Cdk1
Molecular Formula C18H13N3OS2
Formula Weight 351.5g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 5 mg