missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Calbiochem™ BQ-788, Sodium Salt
Highly selective ETB-receptor antagonist
$433.79 - $1,850.00
Specifications
| Color | White to Off-white |
|---|---|
| Content And Storage | -20°C |
| Product Type | BQ-788, Sodium Salt |
| Form | Lyophilized Solid |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
052238385MG
|
MilliporeSigma™
052238385MG |
5 mg |
Each for $1,850.00
|
|
|||||
|
052238381MG
|
MilliporeSigma™
052238381MG |
1 mg |
Each for $433.79
|
|
|||||
Specifications
| White to Off-white | |
| -20°C | |
| BQ-788, Sodium Salt | |
| Lyophilized Solid |
For Research Use Only