missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Benzamidine, Hydrochloride
Inhibitor of trypsin and trypsin-like enzymes
$94.40 - $159.00
Specifications
| Color | White |
|---|---|
| Content And Storage | 15°C to 30°C |
| Product Type | Benzamidine, Hydrochloride |
| Form | Solid |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
1990015GM
|
MilliporeSigma™
1990015GM |
5 g |
Each for $94.40
|
|
|||||
|
19900125GM
|
MilliporeSigma™
19900125GM |
25 g |
Each for $159.00
|
|
|||||
Description
- Inhibits Factor VII autoactivation
Specifications
| White | |
| 15°C to 30°C | |
| Benzamidine, Hydrochloride | |
| Solid |
For Research Use Only