missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ BCIP/NBT Substrates for AP and HRP

Catalog No. 203790100ML
Change view
Click to view available options
Form:
Liquid
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Form
203790100ML Liquid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 203790100ML Supplier MilliporeSigma™ Supplier No. 203790100ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ready-to-use alkaline phosphatase substrate solution. MilliporeSigma™; Calbiochem™ BCIP/NBT Substrates for AP and HRP is suitable for immunoblotting.

  • Form: Clear to pale yellow liquid

Specifications

Kit Size 100mL
Product Type BCIP/NBT Substrates
Formulation 0.577mM BCIP, 0.122mM NBT in a proprietary solution
For Use With (Application) Immunoblotting
Storage Requirements Protect from light. Do not freeze.
Form Liquid