missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Alsterpaullone
Description
- Packaged Under Inert Gas
- Cell Permeable
Specifications
Specifications
| Color | Brown |
| Description | 95% by HPLC |
| Target Proteins | GSK-3β, Cdk1/cyclin B |
| CAS | 237430-03-4 |
| Molecular Formula | C16H11N3O3 |
| Formula Weight | 293.3g/mol |
| Solubility | DMSO |
| Content And Storage | Protect from light |
| Form | Solid |
| Quantity | 1 mg |