missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Adenosine 5'-O-(3-Thiotriphosphate), Tetralithium Salt

Catalog No. 11912025MG
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11912025MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11912025MG Supplier MilliporeSigma™ Supplier No. 11912025MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Used as substrate, inhibitor of ATP-dependent enzyme systems

  • Relatively stable, slowly hydrolyzable ATP analog
  • Can substitute for ATP in various kinase reactions
  • Increases activity of Ca2+-activated K+channels

Specifications

Format White solid
Product Type Adenosine 5′-O-(3-Thiotriphosphate), Tetralithium Salt
Content And Storage -20°C
Purity ≥85% by HPLC
Quantity 25 mg

For Research Use Only