missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbicohem™ Calbiochem™ Catalase Assay Kit

Catalog No. 2192651KIT
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
2192651KIT 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 2192651KIT Supplier MilliporeSigma™ Supplier No. 2192651KIT

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Assay Buffer, Sample Buffer, Formaldehyde Standard, Catalase, Potassium Hydroxide, Methanol, Hydrogen Peroxide, Purpald (Chromogen), Potassium Periodate, 96-well Plate, Plate Cover

Used for measurement of catalase activity in plasma.

EMD Millipore Calbicohem™ Catalase Assay Kit is sensitive spectrophotometric assay.

  • Colorimetric method of detection
  • Measures catalase activity in plasma, serum, erythrocyte lysates, tissue homogenates, cell lysates
  • Assay range: 0.25 to 4nmol/min/mL
  • Contains sufficient reagents for 96 tests
  • Format 96 well plate
  • TotalAssayTime 2 hr
  • AssayRange 0.25 to 4nmol/min/mL

Specifications

Detection Method Colorimetric
Quantity 1 Kit
Content And Storage 2° to 8°C