missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calamine, Powder, USP, 98-100.5%, Spectrum™ Chemical
Description
Spectrum™ Chemical Calamine, Powder, USP is used in topical medications to relieve pain and itching. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
| CAS Max % | 100% |
| Packaging | Poly Bottle |
| Quantity | 125 g |
| Percent Purity | 98-100.5% |
| Grade | USP |