missing translation for 'onlineSavingsMsg'
Learn More

IKA™ CAL-O-50 Silicon Oil

Catalog No. 01257902
Change view
Click to view available options
Product Type:
Standard Silicon Oil
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
01257902 Standard Silicon Oil
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01257902 Supplier IKA™ Supplier No. 0025000399

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Certified standard silicon oil designed to allow you to check the measurement accuracy of your ROTAVISC.

  • Includes certificate
  • For use with a range of ROTAVISC models

Specifications

Product Type Standard Silicon Oil
Viscosity Range 50 cSt Dynamic
Description Period of Validity: 12 months
Temperature Range (Metric) 25°C
Volume (Metric) 500 mL