missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STERIS™ Cage-Klenz 200 Acid-Based Detergent
Description
Cage-Klenz 200 Detergent is specially formulated to remove challenging hard water and urine scales found in research animal caging.
HOW CAGE-KLENZ 200 DETERGENT WORKS
Cage-Klenz 200 Detergent is a phosphoric acid-based detergent for use in high pressure spray animal cage washing equipment.
WHY CAGE-KLENZ 200 DETERGENT?
- Efficacy - Effectively cleans hard water and urine scale, and metal oxides in both soft and hard water
- Flexibility - Excellent materials compatibility with a wide variety of caging materials including polycarbonate
- Cost Savings - Reduces utility consumption and cleaning time due with low foaming, free-rinsing formulation
- Environmentally Friendly - formulation with biodegradable surfactants
- Reliability - Manufactured in a cGMP compliant, FDA registered and ISO 9001 certified facility
Physical Properties: pH (1%): 2.0, typical
Specifications
Specifications
| Product Type | Detergent |
| Color-coded | No |
| Form | Clear Liquid |
| pH | 2.0 (Diluted) |
| Scent | Unscented |
| Sterility | Non-sterile |
| Container Type | Pail |
| Size | 5 gal. |