missing translation for 'onlineSavingsMsg'
Learn More

STERIS™ Cage-Klenz 200 Acid-Based Detergent

Catalog No. 19999639
Change view
Click to view available options
Size:
5 gal.
30 gal.
200 gal.
Container Type:
Drum
Pail
Tote
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Container Type
19999639 5 gal. Pail
19999638 30 gal. Drum
19999640 200 gal. Tote
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19999639 Supplier STERIS™ Supplier No. 1K2005
Only null left

Cage-Klenz 200 Detergent: Phosphoric acid-based formula removes hard water and urine scales in research animal caging. Materials-compatible, cost-saving, and environmentally friendly. pH (1%): 2.0.

Cage-Klenz 200 Detergent is specially formulated to remove challenging hard water and urine scales found in research animal caging.

HOW CAGE-KLENZ 200 DETERGENT WORKS

Cage-Klenz 200 Detergent is a phosphoric acid-based detergent for use in high pressure spray animal cage washing equipment.

WHY CAGE-KLENZ 200 DETERGENT?

  • Efficacy - Effectively cleans hard water and urine scale, and metal oxides in both soft and hard water
  • Flexibility - Excellent materials compatibility with a wide variety of caging materials including polycarbonate
  • Cost Savings - Reduces utility consumption and cleaning time due with low foaming, free-rinsing formulation
  • Environmentally Friendly - formulation with biodegradable surfactants
  • Reliability - Manufactured in a cGMP compliant, FDA registered and ISO 9001 certified facility

Physical Properties: pH (1%): 2.0, typical

Specifications

Product Type Detergent
Color-coded No
Form Clear Liquid
pH 2.0 (Diluted)
Scent Unscented
Sterility Non-sterile
Container Type Pail
Size 5 gal.