Learn More
STERIS Cage-Klenz 100 Alkaline Detergent
Description
- Cage-Klenz 100 Detergent is specially formulated to meet the unique cleaning and materials compatibility requirements found in research animal caging.
- Cage-Klenz 100 Detergent is a liquid, phosphate-free detergent for use in high pressure spray animal cage washing equipment. It is formulated with corrosion inhibitors for enhanced materials compatibility.
- Efficacy - Effectively cleans organic cage residues in both soft and hard water
- Flexibility - Excellent materials compatibility with a wide variety of caging materials including polycarbonate
- Cost Savings - Reduces utility consumption and cleaning time due with low foaming, free-rinsing formulation
- Environmentally Friendly - phosphate-free formulation with biodegradable surfactants
- Reliability - Manufactured in a cGMP compliant, FDA registered and ISO 9001 certified facility
Specifications
Specifications
| Product Type | Detergent |
| Color-coded | No |
| Form | Clear Liquid |
| Scent | Unscented |
| Sterility | Non-sterile |
| Size | 5 gal. |